5-[3-(Trifluoromethoxy)phenyl]nicotinic acid

Lieferant: Apollo Scientific

PC510046-5G
APOSPC510046-5GEA 1540 EUR
APOSPC510046-5G
5-[3-(Trifluoromethoxy)phenyl]nicotinic acid
5-[3-(Trifluoromethoxy)phenyl]nicotinic acid
Formel: C₁₃H₈F₃NO₃
Molecular Weight: 283,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410278
CAS-Nummer: 924854-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR