6-[4-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

Synonyme: 2-(4-(Trifluoromethoxy)phenyl)pyridine-5-carboxaldehyde

PC510047-5G
APOSPC510047-5GEA 1940 EUR
APOSPC510047-5G
6-[4-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde
6-[4-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde
Formel: C₁₃H₈F₃NO₂
Molecular Weight: 267,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115436
CAS-Nummer: 851069-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR