3-(Boc-amino)-3-(4-fluorophenyl)-N-hydroxypropanamide
Lieferant: Apollo Scientific
PC510048-5G
APOSPC510048-5GEA
1330
EUR
APOSPC510048-5G
3-(Boc-amino)-3-(4-fluorophenyl)-N-hydroxypropanamide
3-(Boc-amino)-3-(4-fluorophenyl)-N-hydroxypropanamide
Formel:
C₁₄H₁₉FN₂O₄ Molecular Weight: 298,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115558 CAS-Nummer: 898404-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...