4-[(Cyclopropylcarbonyl)amino]-2-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific

PC510052-1G PC510052-5G
APOSPC510052-1GEA 306 EUR
APOSPC510052-1G APOSPC510052-5G
4-[(Cyclopropylcarbonyl)amino]-2-(trifluoromethyl)benzoic acid
4-[(Cyclopropylcarbonyl)amino]-2-(trifluoromethyl)benzoic acid
Formel: C₁₂H₁₀F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19442790
CAS-Nummer: 1314406-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR