1-(Boc-amino)-3-bromo-1-(3-fluorophenyl)propane

Lieferant: Apollo Scientific

PC510063-5G
APOSPC510063-5GEA 1620 EUR
APOSPC510063-5G
1-(Boc-amino)-3-bromo-1-(3-fluorophenyl)propane
1-(Boc-amino)-3-bromo-1-(3-fluorophenyl)propane
Formel: C₁₄H₁₉BrFNO₂
Molecular Weight: 332.209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410614
CAS-Nummer: 924817-99-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR