5-[4-(Trifluoromethyl)phenyl]-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

PC510064-5G
APOSPC510064-5GEA 1840 EUR
APOSPC510064-5G
5-[4-(Trifluoromethyl)phenyl]-3-pyridinecarbaldehyde
5-[4-(Trifluoromethyl)phenyl]-3-pyridinecarbaldehyde
Formel: C₁₃H₈F₃NO
Molecular Weight: 251,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410235
CAS-Nummer: 885959-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR