6-[3-(Trifluoromethoxy)phenyl]-2-pyridinecarbaldehyde

Lieferant: Apollo Scientific

PC510070-5G
APOSPC510070-5GEA 1840 EUR
APOSPC510070-5G
6-[3-(Trifluoromethoxy)phenyl]-2-pyridinecarbaldehyde
6-[3-(Trifluoromethoxy)phenyl]-2-pyridinecarbaldehyde
Formel: C₁₃H₈F₃NO₂
Molecular Weight: 267,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410030
CAS-Nummer: 887980-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR