6-(4-Fluorophenyl)nicotinic acid

Lieferant: Apollo Scientific

PC510071-5G
APOSPC510071-5GEA 1540 EUR
APOSPC510071-5G
6-(4-Fluorophenyl)nicotinic acid
6-(4-Fluorophenyl)nicotinic acid
Formel: C₁₂H₈FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06410388
CAS-Nummer: 223127-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR