[6-[4-(Trifluoromethoxy)phenyl]-3-pyridyl]methanol

Lieferant: Apollo Scientific

Synonyme: 2-(4-(Trifluoromethoxy)phenyl)pyridine-5-methanol

PC510072-5G
APOSPC510072-5GEA 1840 EUR
APOSPC510072-5G
[6-[4-(Trifluoromethoxy)phenyl]-3-pyridyl]methanol
[6-[4-(Trifluoromethoxy)phenyl]-3-pyridyl]methanol
Formel: C₁₃H₁₀F₃NO₂
Molecular Weight: 269,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410451
CAS-Nummer: 851069-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR