6-[3-(Trifluoromethoxy)phenyl]nicotinic acid

Lieferant: Apollo Scientific

PC510075-5G
APOSPC510075-5GEA 1840 EUR
APOSPC510075-5G
6-[3-(Trifluoromethoxy)phenyl]nicotinic acid
6-[3-(Trifluoromethoxy)phenyl]nicotinic acid
Formel: C₁₃H₈F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06410407
CAS-Nummer: 197847-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR