[6-[3-(Trifluoromethyl)phenyl]-3-pyridyl]methanol

Lieferant: Apollo Scientific

PC510076-5G
APOSPC510076-5GEA 1840 EUR
APOSPC510076-5G
[6-[3-(Trifluoromethyl)phenyl]-3-pyridyl]methanol
[6-[3-(Trifluoromethyl)phenyl]-3-pyridyl]methanol
Formel: C₁₃H₁₀F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06410447
CAS-Nummer: 887974-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR