1-[8-(4-Fluorophenyl)-1,4-dioxaspiro[4.5]dec-8-yl]methanamine

Lieferant: Apollo Scientific

PC510080-1G
APOSPC510080-1GEA 713 EUR
APOSPC510080-1G
1-[8-(4-Fluorophenyl)-1,4-dioxaspiro[4.5]dec-8-yl]methanamine
1-[8-(4-Fluorophenyl)-1,4-dioxaspiro[4.5]dec-8-yl]methanamine
Formel: C₁₅H₂₀FNO₂
Molecular Weight: 265,323 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410588
CAS-Nummer: 887979-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR