Methyl 4-(4-benzyl-1-piperazinyl)-3-fluorobenzoate
Lieferant: Apollo Scientific
PC510082-5G
APOSPC510082-5GEA
601
EUR
APOSPC510082-5G
Methyl 4-(4-benzyl-1-piperazinyl)-3-fluorobenzoate
Methyl 4-(4-benzyl-1-piperazinyl)-3-fluorobenzoate
Formel:
C₁₉H₂₁FN₂O₂ Molecular Weight: 328,381 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134208 CAS-Nummer: 1346597-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...