4-(4-Benzyl-1-piperazinyl)-3-fluorobenzoic acid
Lieferant: Apollo Scientific
PC510083-5G
APOSPC510083-5GEA
713
EUR
APOSPC510083-5G
4-(4-Benzyl-1-piperazinyl)-3-fluorobenzoic acid
4-(4-Benzyl-1-piperazinyl)-3-fluorobenzoic acid
Formel:
C₁₈H₁₉FN₂O₂ Molecular Weight: 314,354 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134209 CAS-Nummer: 1346597-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...