Methyl 3-Fluoro-4-(4-methyl-1-piperazinyl)benzoate
Lieferant: Apollo Scientific
PC510084-5G
APOSPC510084-5GEA
601
EUR
APOSPC510084-5G
Methyl 3-Fluoro-4-(4-methyl-1-piperazinyl)benzoate
Methyl 3-Fluoro-4-(4-methyl-1-piperazinyl)benzoate
Formel:
C₁₃H₁₇FN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134210 CAS-Nummer: 948018-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...