3-[Benzyl(methyl)amino]-2,2-difluoropropanoic acid

Lieferant: Apollo Scientific

PC510087-1G
APOSPC510087-1GEA 713 EUR
APOSPC510087-1G
3-[Benzyl(methyl)amino]-2,2-difluoropropanoic acid
3-[Benzyl(methyl)amino]-2,2-difluoropropanoic acid
Formel: C₁₁H₁₃F₂NO₂
Molecular Weight: 229,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20134215
CAS-Nummer: 1346597-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR