Ethyl 3-[Benzyl(methyl)amino]-2,2-difluoropropanoate
Lieferant: Apollo Scientific
PC510088-1G
APOSPC510088-1GEA
601
EUR
APOSPC510088-1G
Ethyl 3-[Benzyl(methyl)amino]-2,2-difluoropropanoate
Ethyl 3-[Benzyl(methyl)amino]-2,2-difluoropropanoate
Formel:
C₁₃H₁₇F₂NO₂ Molecular Weight: 257,276 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134216 CAS-Nummer: 1346597-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...