6-[2-(Trifluoromethoxy)phenyl]-2-pyridinecarboxylic acid

Lieferant: Apollo Scientific

PC510089-1G PC510089-5G
APOSPC510089-1GEA 448 EUR
APOSPC510089-1G APOSPC510089-5G
6-[2-(Trifluoromethoxy)phenyl]-2-pyridinecarboxylic acid
6-[2-(Trifluoromethoxy)phenyl]-2-pyridinecarboxylic acid
Formel: C₁₃H₈F₃NO₃
Molecular Weight: 283.203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410072
CAS-Nummer: 887983-48-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR