5-(2-Fluorophenyl)-2-thiophenecarbaldehyde

Lieferant: Apollo Scientific

PC510091-5G
APOSPC510091-5GEA 1120 EUR
APOSPC510091-5G
5-(2-Fluorophenyl)-2-thiophenecarbaldehyde
5-(2-Fluorophenyl)-2-thiophenecarbaldehyde
Formel: C₁₁H₇FOS
Molecular Weight: 206,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410179
CAS-Nummer: 886508-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR