5-(2-Fluorphenyl)-3-pyridincarbaldehyd

Lieferant: Apollo Scientific

PC510093-5G
APOSPC510093-5GEA 1620 EUR
APOSPC510093-5G
5-(2-Fluorphenyl)-3-pyridincarbaldehyd
5-(2-Fluorphenyl)-3-pyridincarbaldehyd
Formel: C₁₂H₈FNO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 887973-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR