6-[(4-Trifluoromethyl)phenyl]nicotinaldehyde

Lieferant: Apollo Scientific

PC510095-1G
APOSPC510095-1GEA 499 EUR
APOSPC510095-1G
6-[(4-Trifluoromethyl)phenyl]nicotinaldehyde
6-[(4-Trifluoromethyl)phenyl]nicotinaldehyde
Formel: C₁₃H₈F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD04115430
CAS-Nummer: 356058-14-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR