1-Boc-4-(5-bromo-3-fluoro-2-pyridyl)-3-methylpiperazine

Lieferant: Apollo Scientific

PC510098-1G
APOSPC510098-1GEA 916 EUR
APOSPC510098-1G
1-Boc-4-(5-bromo-3-fluoro-2-pyridyl)-3-methylpiperazine
1-Boc-4-(5-bromo-3-fluoro-2-pyridyl)-3-methylpiperazine
Formel: C₁₅H₂₁BrFN₃O₂
Molecular Weight: 374,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21332888
CAS-Nummer: 1354940-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR