6-Bromo-7-fluoro-3,4-dihydroquinolin-2(1H)-one

Lieferant: Apollo Scientific

PC510099-1G
APOSPC510099-1GEA 713 EUR
APOSPC510099-1G
6-Bromo-7-fluoro-3,4-dihydroquinolin-2(1H)-one
6-Bromo-7-fluoro-3,4-dihydroquinolin-2(1H)-one
Formel: C₉H₇BrFNO
Molecular Weight: 244,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12174779
CAS-Nummer: 1156389-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR