2-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,3-dioxolane

Lieferant: Apollo Scientific

PC510100-5G
APOSPC510100-5GEA 601 EUR
APOSPC510100-5G
2-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,3-dioxolane
2-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,3-dioxolane
Formel: C₁₀H₁₁FO₄S
Molecular Weight: 246,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20491403
CAS-Nummer: 1346597-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR