2,2-Difluoro-3-(methylamino)propanoic acid hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
PC510102-1G
APOSPC510102-1GEA
804
EUR
APOSPC510102-1G
2,2-Difluoro-3-(methylamino)propanoic acid hydrochloride
2,2-Difluoro-3-(methylamino)propanoic acid hydrochloride
Formel:
C₄H₇F₂NO₂·ClH Molecular Weight: 175,562 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20491408 CAS-Nummer: 1346597-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...