2-[4-Fluoro-3-(methylsulphonyl)phenyl]-1,3-dioxolane
Lieferant: Apollo Scientific
PC510105-5G
APOSPC510105-5GEA
601
EUR
APOSPC510105-5G
2-[4-Fluoro-3-(methylsulphonyl)phenyl]-1,3-dioxolane
2-[4-Fluoro-3-(methylsulphonyl)phenyl]-1,3-dioxolane
Formel:
C₁₀H₁₁FO₄S Molecular Weight: 246,255 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21332896 CAS-Nummer: 1354940-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...