Methyl-4-bromo-2-fluorophenylacetate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(4-bromo-2-fluorophenyl)acetate

PC510106-5G
APOSPC510106-5GEA 55.1 EUR
APOSPC510106-5G
Methyl-4-bromo-2-fluorophenylacetate
Methyl-4-bromo-2-fluorophenylacetate
Formel: C₉H₈BrFO₂
Molecular Weight: 247.063 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20483545
CAS-Nummer: 193290-19-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR