Ethyl 1-Cbz-3-fluoropiperidine-3-carboxylate

Lieferant: Apollo Scientific

PC510108-5G
APOSPC510108-5GEA 916 EUR
APOSPC510108-5G
Ethyl 1-Cbz-3-fluoropiperidine-3-carboxylate
Ethyl 1-Cbz-3-fluoropiperidine-3-carboxylate
Formel: C₁₆H₂₀FNO₄
Molecular Weight: 309,333 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641307
CAS-Nummer: 1111640-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR