Methyl 1-Cbz-3-fluoropiperidine-3-carboxylate

Lieferant: Apollo Scientific

PC510109-5G
APOSPC510109-5GEA 916 EUR
APOSPC510109-5G
Methyl 1-Cbz-3-fluoropiperidine-3-carboxylate
Methyl 1-Cbz-3-fluoropiperidine-3-carboxylate
Formel: C₁₅H₁₈FNO₄
Molecular Weight: 295,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641308
CAS-Nummer: 1363166-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR