1-Cbz-3-fluoropiperidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC510110-5G
APOSPC510110-5GEA 1210 EUR
APOSPC510110-5G
1-Cbz-3-fluoropiperidine-3-carboxylic acid
1-Cbz-3-fluoropiperidine-3-carboxylic acid
Formel: C₁₄H₁₆FNO₄
Molecular Weight: 281,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641309
CAS-Nummer: 1363166-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR