Potassium Trifluoro(4-methoxybenzyl)borate
Lieferant: Apollo Scientific
Synonyme:
Potassium trifluoro(4-methoxybenzyl)borate
PC510113-1G
APOSPC510113-1GEA
713
EUR
APOSPC510113-1G
Potassium Trifluoro(4-methoxybenzyl)borate
Potassium Trifluoro(4-methoxybenzyl)borate
Formel:
C₈H₉BF₃O·K Molecular Weight: 228,061 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09993297 CAS-Nummer: 900810-91-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...