1-Acetyl-3-bromo-6-fluoroindole
Lieferant: Apollo Scientific
PC510115-5G
APOSPC510115-5GEA
510
EUR
APOSPC510115-5G
1-Acetyl-3-bromo-6-fluoroindole
1-Acetyl-3-bromo-6-fluoroindole
Formel:
C₁₀H₇BrFNO Molecular Weight: 256,071 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22205867 CAS-Nummer: 1375064-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...