Ethyl 3-Bromo-6-fluoroindole-1-carboxylate

Lieferant: Apollo Scientific

PC510116-5G
APOSPC510116-5GEA 510 EUR
APOSPC510116-5G
Ethyl 3-Bromo-6-fluoroindole-1-carboxylate
Ethyl 3-Bromo-6-fluoroindole-1-carboxylate
Formel: C₁₁H₉BrFNO₂
Molecular Weight: 286,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205868
CAS-Nummer: 1375064-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR