1-Acetyl-3-bromo-5-fluoroindole
Lieferant: Apollo Scientific
PC510117-5G
APOSPC510117-5GEA
713
EUR
APOSPC510117-5G
1-Acetyl-3-bromo-5-fluoroindole
1-Acetyl-3-bromo-5-fluoroindole
Formel:
C₁₀H₇BrFNO Molecular Weight: 256,071 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22205879 CAS-Nummer: 1375064-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...