Ethyl 3-Bromo-5-fluoroindole-1-carboxylate

Lieferant: Apollo Scientific

PC510118-5G
APOSPC510118-5GEA 713 EUR
APOSPC510118-5G
Ethyl 3-Bromo-5-fluoroindole-1-carboxylate
Ethyl 3-Bromo-5-fluoroindole-1-carboxylate
Formel: C₁₁H₉BrFNO₂
Molecular Weight: 286.097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205880
CAS-Nummer: 1375064-48-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR