1-(3-Bromo-4-fluorophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

PC510119-5G
APOSPC510119-5GEA 1220 EUR
APOSPC510119-5G
1-(3-Bromo-4-fluorophenyl)-4-oxocyclohexanecarboxylic acid
1-(3-Bromo-4-fluorophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₂BrFO₃
Molecular Weight: 315,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423118
CAS-Nummer: 1385694-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR