6-Bromo-5-fluoro-1-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC51012-250MG
APOSPC51012-250MGEA 72.5 EUR
APOSPC51012-250MG
6-Bromo-5-fluoro-1-methyl-1H-indazole
6-Bromo-5-fluoro-1-methyl-1H-indazole
Formel: C₈H₆BrFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18089817
CAS-Nummer: 1286734-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR