Methyl 1-(3-Bromo-4-fluorophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

PC510120-5G
APOSPC510120-5GEA 1220 EUR
APOSPC510120-5G
Methyl 1-(3-Bromo-4-fluorophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(3-Bromo-4-fluorophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₄H₁₄BrFO₃
Molecular Weight: 329,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423121
CAS-Nummer: 1385694-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR