4-Oxo-1-[3-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid

Lieferant: Apollo Scientific

PC510121-5G
APOSPC510121-5GEA 1220 EUR
APOSPC510121-5G
4-Oxo-1-[3-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid
4-Oxo-1-[3-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid
Formel: C₁₄H₁₃F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22423124
CAS-Nummer: 1385694-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR