Methyl 4-Oxo-1-[3-(trifluoromethyl)phenyl]cyclohexanecarboxylate

Lieferant: Apollo Scientific

PC510122-5G
APOSPC510122-5GEA 1220 EUR
APOSPC510122-5G
Methyl 4-Oxo-1-[3-(trifluoromethyl)phenyl]cyclohexanecarboxylate
Methyl 4-Oxo-1-[3-(trifluoromethyl)phenyl]cyclohexanecarboxylate
Formel: C₁₅H₁₅F₃O₃
Molecular Weight: 300,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423125
CAS-Nummer: 1385694-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR