1-(3-Fluoro-4-methoxyphenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

PC510123-5G
APOSPC510123-5GEA 1220 EUR
APOSPC510123-5G
1-(3-Fluoro-4-methoxyphenyl)-4-oxocyclohexanecarboxylic acid
1-(3-Fluoro-4-methoxyphenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₄H₁₅FO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22423126
CAS-Nummer: 1385694-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR