Methyl 1-(3-Fluoro-4-methoxyphenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

PC510124-5G
APOSPC510124-5GEA 1220 EUR
APOSPC510124-5G
Methyl 1-(3-Fluoro-4-methoxyphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(3-Fluoro-4-methoxyphenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₅H₁₇FO₄
Molecular Weight: 280,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423127
CAS-Nummer: 1385694-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR