1-(3-Fluorophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

PC510125-5G
APOSPC510125-5GEA 1010 EUR
APOSPC510125-5G
1-(3-Fluorophenyl)-4-oxocyclohexanecarboxylic acid
1-(3-Fluorophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₃FO₃
Molecular Weight: 236,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17013938
CAS-Nummer: 374795-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR