Methyl 1-(3-Fluorophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

PC510126-5G
APOSPC510126-5GEA 1010 EUR
APOSPC510126-5G
Methyl 1-(3-Fluorophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(3-Fluorophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₄H₁₅FO₃
Molecular Weight: 250,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423131
CAS-Nummer: 1384264-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR