4-Oxo-1-[4-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid

Lieferant: Apollo Scientific

PC510127-5G
APOSPC510127-5GEA 1120 EUR
APOSPC510127-5G
4-Oxo-1-[4-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid
4-Oxo-1-[4-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid
Formel: C₁₄H₁₃F₃O₃
Molecular Weight: 286,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423132
CAS-Nummer: 1385694-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR