Methyl 4-Oxo-1-(4-(trifluoromethyl)phenyl)cyclohexanecarboxylate

Lieferant: Apollo Scientific

PC510128-5G
APOSPC510128-5GEA 1120 EUR
APOSPC510128-5G
Methyl 4-Oxo-1-(4-(trifluoromethyl)phenyl)cyclohexanecarboxylate
Methyl 4-Oxo-1-(4-(trifluoromethyl)phenyl)cyclohexanecarboxylate
Formel: C₁₅H₁₅F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22423133
CAS-Nummer: 1242314-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR