Methyl 4-Oxo-1-(2,4,5-trifluorophenyl)cyclohexanecarboxylate

Lieferant: Apollo Scientific

PC510129-5G
APOSPC510129-5GEA 1010 EUR
APOSPC510129-5G
Methyl 4-Oxo-1-(2,4,5-trifluorophenyl)cyclohexanecarboxylate
Methyl 4-Oxo-1-(2,4,5-trifluorophenyl)cyclohexanecarboxylate
Formel: C₁₄H₁₃F₃O₃
Molecular Weight: 286,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423134
CAS-Nummer: 1385694-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR