4-Oxo-1-(2,4,5-trifluorophenyl)cyclohexanecarboxylic acid

Lieferant: Apollo Scientific

PC510130-5G
APOSPC510130-5GEA 1010 EUR
APOSPC510130-5G
4-Oxo-1-(2,4,5-trifluorophenyl)cyclohexanecarboxylic acid
4-Oxo-1-(2,4,5-trifluorophenyl)cyclohexanecarboxylic acid
Formel: C₁₃H₁₁F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22423135
CAS-Nummer: 1385694-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR