(2-Fluorophenyl)trimethylsilane

Lieferant: Apollo Scientific

PC510135-5G
APOSPC510135-5GEA 205 EUR
APOSPC510135-5G
(2-Fluorophenyl)trimethylsilane
(2-Fluorophenyl)trimethylsilane
Formel: C₉H₁₃FSi
Lagertemperatur: Raumtemperatur
MDL: MFCD00092633
CAS-Nummer: 1842-26-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR