5-Bromo-3-(trifluoroacetyl)indole
Lieferant: Apollo Scientific
Synonyme:
1-(5-Bromo-1H-indol-3-yl)-2,2,2-trifluoroethanone
PC510142-5G
PC510142-1G
APOSPC510142-5GEA
397
EUR
APOSPC510142-5G
APOSPC510142-1G
5-Bromo-3-(trifluoroacetyl)indole
5-Bromo-3-(trifluoroacetyl)indole
Formel:
C₁₀H₅BrF₃NO Molecular Weight: 292.052 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03721051 CAS-Nummer: 32387-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...